Celite® 545, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
3376.2500
: Avantor
:
- Pakning:Glass bottle for solids
- Pakn:2,5 kg
- Seq:4076
- Environmentally Preferable:
- Loss on Drying at 110°C:max. 0.2 %
- Petroleum Ether Extractables:value on label
- pH of 5% Slurry at 20°C:8.0 - 11.0
: DE, Diatomaceous earth, flux-calcined
:
:60.08 g/mol
:272-489-0
:GHS08
:68855-54-9
:SiO₂
:MFCD00132803