Orders LinkContactUsLinkComponent
Collagenase B, ACF
Collagenase B, ACF
Marketsource
Leverandør:   Stemcell Technologies
Collagenase B, ACF
Leverandør:  Stemcell Technologies
Marketsource
Certificates
Collagenase B, ACF
2 Options Available Below
Product Details & Documents
Animal component-free collagenase for the digestion of native collagen fibrils.
  • Obtain cleaner cultures with an animal component-free solution
Use Collagenase B, Animal Component-Free (ACF) to digest native collagen fibrils in connective tissues and prevent the introduction of animal-derived pathogens. This enzymatic preparation is sourced from Clostridium histolyticum and is obtained from cultures free of animal-derived materials. Collagenase preparations contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler and Yaron). Collagenase B, ACF contains higher collagenase and caseinase levels than Collagenase A.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • STMC07438
  • STMC07439
  • Species:
  • Non-human primate|Human|Rat|Mouse
  • Non-human primate|Human|Rat|Mouse
Specifications
Product Family Options
Product Information
  • Pakn
    Tilgængelighed
    Price
  • STMC07438
    Collagenase B, ACF
    Collagenase B, ACF
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 1.233,00 kr
    1.233,00 kr
    1 * 100 mg
       1.233,00 kr
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC07438
    Species
    Non-human primate|Human|Rat|Mouse
    Marketsource
  • STMC07439
    Collagenase B, ACF
    Collagenase B, ACF
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 5.651,00 kr
    5.651,00 kr
    1 * 1 g
       5.651,00 kr
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC07439
    Species
    Non-human primate|Human|Rat|Mouse