Orders LinkContactUsLinkComponent
CellAdhere™ Vitronectin, Human, Solution
CellAdhere™ Vitronectin, Human, Solution
CellAdhere™ Vitronectin, human solution, Flydende, 0,1 mg
Katalog # STMC07004
Marketsource
Leverandør:   Stemcell Technologies
CellAdhere™ Vitronectin, Human, Solution
Katalog # STMC07004
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
CellAdhere™ Vitronectin, Human, Solution
CellAdhere™ Vitronectin, Human, Solution
CellAdhere™ Vitronectin, human solution, Flydende, 0,1 mg
CAS nummer:  
Price
3.267,00 kr
Log ind for tilgængelighed
1 * 0,1 mg
Product Details & Documents
Purified human vitronectin solution for tissue engineering research and cell culture - 0,2 ml.
Use CellAdhere™ Vitronectin, human, solution as a cultureware coating to promote the attachment and spreading of cell cultures. This glycoprotein surface coating is purified from human serum by the method of Hayman et al. using an anti-Vitronectin monoclonal antibody affinity column and follow by sterilisation with 0,2 µm filtration. Vitronectin has a purity of >95% based on Coomassie brilliant blue stain of 7,5% SDS-PAGE. This product is dissolved in 0,2 ml of 0,15 M NaCl, 0,005 M HEPES buffer at an approximate pH 7,4. The final concentration of Vitronectin is 0,5 mg/ml. Fibronectin contamination is less than 0,04% based on immunoblotting. The optimal protein concentration may vary depending on the cell type being used and therefore must be titrated for best results.
Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet ( collectively, the 'Product Restrictions').
Specifikationer
Specifikationer
  • Medieformat:
    Flydende
  • Pakn:
    0,1 mg
  • Beskrivelse:
    CellAdhere™ Vitronectin, human solution