KingFisher™ Presto Purification System
KingFisher™ Presto with 96 DW head, 96 samples per run, 5...115 °C heating tempertarure
Katalog # FINN5400830
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Use the Thermo Scientific KingFisher Presto as part of an automated purification system with a liquid handler for plate filling in high-throughput laboratories.
- Volume range of 50 to 5000 μl
- Minimised size for easy fitting to automation systems
- Heating capability for application requiring elevated temperatures
- Easy connection to several liquid handling instruments
- Flexible system for wide variety of applications
Utilising proven KingFisher magnetic particle-based technology, the KingFisher Presto provides high-quality yields of target nucleic acids and proteins for biopharma, biotech and research.
Specifikationer
- Beskrivelse:KingFisher™ Presto with 96 DW head, 96 samples per run, 5...115 °C heating tempertarure
- Cat. No.:FINN5400830
- B×D×H:360×465×400 mm
- Vægt:24 kg
- Behandlingsvolumen:50 to 1000 μl
- Pladetyper:96, 96 DW