Silicone Hose
Silicone hose, vacuum-resistant, 6 mm, 3 mm, 12 mm, transparent
Katalog # NEUB6201-060-030
Leverandør: NEUBERT VOLUME GLASSWAERE
Gå til:
Product Details & Documents
Hose made of quality silicone rubber (VMQ).
- Hardness 55 Shore A
- Food safe
- Peristaltic pumps suitable
Temperature resistant from –60 to +200 °C, short-term up to 260 °C.
Specifikationer
- Rør udv.Ø:12 mm
- Rør int.Ø:6 mm
- Vægtykkelse:3 mm
- Farve:transparent
- Beskrivelse:Silicone hose, vacuum-resistant
- Seq:01