Orders LinkContactUsLinkComponent
EasySep™ Release Mouse APC Positive Selection Kit
EasySep™ Release Mouse APC Positive Selection Kit
EasySep™ Release Mouse APC positive selection kit, 1×10⁹ cells
Katalog # STMC100-0033
Marketsource
Leverandør:   Stemcell Technologies
EasySep™ Release Mouse APC Positive Selection Kit
Katalog # STMC100-0033
Leverandør:  Stemcell Technologies
Marketsource
EasySep™ Release Mouse APC Positive Selection Kit
EasySep™ Release Mouse APC Positive Selection Kit
EasySep™ Release Mouse APC positive selection kit, 1×10⁹ cells
Price
6.636,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Immunomagnetic positive selection of particle-free mouse cells labeled with APC-conjugated antibodies.
  • Highly purified cells labelled with APC-conjugated antibodies isolated from mouse tissue in less than 40 minutes
  • No-wash removal of EasySep™ Releasable RapidSpheres™
The EasySep™ Release Mouse APC Positive Selection Kit is designed to isolate cells that are labeled with APC-conjugated antibodies from mouse tissue by immunomagnetic positive selection.
Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Then, bound magnetic particles are removed from the EasySep™-isolated, APC-antibody labeled cells, which are immediately available for downstream applications. Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands.
Advarsel: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).
Specifikationer
  • Beskrivelse:
    EasySep™ Release Mouse APC positive selection kit, 1×10⁹ cells