RosetteSep™ Human Custom Cocktail
Product Details & Documents
Immunodensity isolation of untouched human cells.
- Fast and easy-to-use
- Requires no special equipment or training
- Untouched, viable cells
- Can be combined with SepMate™ for consistent, high-throughput sample processing
The RosetteSep™ Human Custom Cocktail is designed to deplete unwanted cells from fresh whole blood by negative selection. When centrifuged over a buoyant density medium such as Lymphoprep™ for more information.
: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).
Specifikationer
- Beskrivelse:RosetteSep™ human Custom cocktail, 1 custom