Remdesivir
Glass bottle, 100 mg
Katalog # 469411000.
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: 2-Ethylbutyl ((S)-(((2R,3S,4R,5R)-5-(4-aminopyrrolo[2,1-f][1,2,4]triazin-7-yl)-5-cyano-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(phenoxy)phosphoryl)-L-alaninate
- CAS nummer:1809249-37-3
- Formel:C₂₇H₃₅N₆O₈P
- MDL nummer:MFCD31657351
- Molecular Weight:602.58 g/mol
- Opbevaringstemperatur:Fryser
Specifikationer
Specifikationstestresultater
- Appearance:white to almost white crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:100 mg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



