Marbofloxacin
Marbofloxacin, 100 mg
Katalog # ACSDSB39230.100MG
Leverandør: CPACHEM
Gå til:
Product Details & Documents
Certifications: ISO 17034, 17025, 9001 certified.
Specifikationer
Specifikationer
- Pakn:100 mg
- Beskrivelse:Marbofloxacin
- Cat. No.:ACSDSB39230.100MG
- CAS-nr:115550-35-1
- Shelf life:24 Months
- Opbevaringstemperatur:Store in a refrigerator at temperatures between 2...8 °C