Orders LinkContactUsLinkComponent
1,1-Dichlorpropan
1,1-Dichlorpropan
100 mg
  ACSDSB23070.100MG
 :   CPACHEM
1,1-Dichlorpropan
  ACSDSB23070.100MG
 :  CPACHEM
 :  78-99-9
1,1-Dichlorpropan
100 mg
 :  78-99-9
1.850,00 kr
1 * 100 mg
Ordering information: Shelf life on ship date: 24 months - store in a refrigerator at temperatures between 2 to 8 °C.
  •  :78-99-9
  •  :Cl₂HCCH₂CH₃
  •  :112.99 g/mol
  •  :88 °C
  •  :7 °C
  •  :Køleskab
  •  :201-165-3
  •  :1993
  •  :3,II
  •  :Density: 1.143 g/cm³ (10 °C)
  • Pakn:
    100 mg