Oven Sample Processor, 874
Oven sample processor for 6 ml sample vessels
Katalog # METR28740120
Leverandør: METROHM
Gå til:
Product Details & Documents
The 874 Oven Sample Processor is used for automatic thermal sample preparation in Karl Fischer titration.
- Oven sample changer: Automated sample preparation for samples that cannot be titrated directly
The oven method is particularly suitable for samples that do not release their water until higher temperatures have been reached, for sparingly soluble samples, or those that react with the KF reagent.
Specifikationer
- Beskrivelse:Oven sample processor for 6 ml sample vessels