BD® Arterial Cannula
BD® Arterial cannula, G 20, 49 ml/min, 45 mm, +
Katalog # BDAM682245
Leverandør: BD Medical
Gå til:
Product Details & Documents
Designed specifically for arterial cannulation, the BD® arterial cannula enables arterial pressure monitoring and serial arterial blood gas determinations.
- Developed to support smooth skin and vein penetration
- Clear visibility of blood flashback confirms arterial entry
- Flexible wings stabilise the catheter in place, helping to prevent unwanted movement
Device is designed to prevent back-flow, which potentially reduces the risk of air emboli and exposure to blood.
Specifikationer
- Flowhastighed:49 ml/min
- Måler:G 20
- Steril:+
- Beskrivelse:BD® Arterial cannula
- Længde:45 mm