Orders LinkContactUsLinkComponent
Axygen® LTS Compatible Pipette Tips
Axygen® LTS Compatible Pipette Tips
   Sustainable Options Available
Supplier:   Corning
Axygen® LTS Compatible Pipette Tips
Supplier:  Corning
   Sustainable Options Available
Certificates
Axygen® LTS Compatible Pipette Tips
16 Options Available Below
Product Details & Documents
The new Axygen® LTS compatible pipette tips are ergonomically designed to reduce discomfort and are optimised to work with Rainin LiteTouch™ pipettors. These tips maintain the quality users have come to expect from corning.
  • Graduation lines for quick accuracy checks
  • Ultra-clear USP class VI resin
  • Fine point delivery system ensures crisp sample dispensing
Flexible sealing area optimised to work with LTS pipettors enables less force to be used during pipetting.
Certifications: ISO 13485, ISO 14001. RNase-free, DNase-free, human DNA-free, PCR inhibitor-free, and non-pyrogenic.
Specifications for Product Family
Specifications
  • Catalog No:
  • 613-7421
  • 613-7422
  • 613-7413
  • 613-7416
  • 613-7410
  • 613-7412
  • 613-7415
  • 613-7409
  • 613-7414
  • 613-7417
  • 613-7411
  • 613-7419
  • 613-7418
  • 613-7424
  • 613-7425
  • 613-7423
  • Environmentally Preferable:
  • Lav fremstillingspåvirkning|Bæredygtig emballage
  • Lav fremstillingspåvirkning|Bæredygtig emballage
  • Lav fremstillingspåvirkning|Bæredygtig emballage
  • Miljøcertificering:
  • Compared to conventional plastic racked pipet tips, reduces plastic per case by 70%.
    Produced in ISO 14001 Certified facility.
  • Compared to conventional plastic racked pipet tips, reduces plastic per case by 70%.
    Produced in ISO 14001 Certified facility.
  • Compared to conventional plastic racked pipet tips, reduces plastic per case by 70%.
    Produced in ISO 14001 Certified facility.
Specifications
Product Family Options
Racked, filtered, sterile
Product Information
  • Spidstype
    Volumen
    Steril
    Filter
    Længde
    Spits farge
    Pakket
    Availability
    Price
  • LTS compatible, extended length
    20 µl
    +
    +
    49,45 mm
    Clear
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 Items
    1 * 960 Items - 830,00 kr
    830,00 kr
    1 * 960 Items
       830,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    613-7421
    Environmentally Preferable
    Miljøcertificering
  • LTS compatible
    200 µl
    +
    +
    50,29 mm
    Clear
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 Items
    1 * 960 Items - 830,00 kr
    830,00 kr
    1 * 960 Items
       830,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    613-7422
    Environmentally Preferable
    Miljøcertificering
Racked, non-filtered, sterile
Product Information
  • Spidstype
    Volumen
    Steril
    Filter
    Længde
    Spits farge
    Pakket
    Availability
    Price
  • LTS compatible, extended length
    20 µl
    +
    -
    49,45 mm
    Clear
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 Items
    1 * 960 Items - 498,00 kr
    498,00 kr
    1 * 960 Items
       498,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    613-7413
    Environmentally Preferable
    Miljøcertificering
  • LTS compatible
    200 µl
    +
    -
    50,29 mm
    Clear
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 Items
    1 * 960 Items - 498,00 kr
    498,00 kr
    1 * 960 Items
       498,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    613-7416
    Environmentally Preferable
    Miljøcertificering
  • LTS compatible, extended length
    1000 µl
    +
    -
    98,58 mm
    Clear
    8 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 768 Items
    1 * 768 Items - 544,00 kr
    544,00 kr
    1 * 768 Items
       544,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    613-7410
    Environmentally Preferable
    Miljøcertificering
Racked, non-filtered, non sterile
Product Information
  • Spidstype
    Volumen
    Steril
    Filter
    Længde
    Spits farge
    Pakket
    Availability
    Price
  • LTS compatible, extended length
    20 µl
    -
    -
    49,45 mm
    Clear
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 Items
    1 * 960 Items - 433,00 kr
    433,00 kr
    1 * 960 Items
       433,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    613-7412
    Environmentally Preferable
    Miljøcertificering
  • LTS compatible
    200 µl
    -
    -
    50,29 mm
    Clear
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 Items
    1 * 960 Items - 473,00 kr
    473,00 kr
    1 * 960 Items
       473,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    613-7415
    Environmentally Preferable
    Miljøcertificering
  • LTS compatible, extended length
    1000 µl
    -
    -
    98,58 mm
    Clear
    8 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 768 Items
    1 * 768 Items - 473,00 kr
    473,00 kr
    1 * 768 Items
       473,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    613-7409
    Environmentally Preferable
    Miljøcertificering
Reload, non-filtered, non sterile
Product Information
  • Spidstype
    Volumen
    Steril
    Filter
    Længde
    Spits farge
    Pakket
    Availability
    Price
  • LTS compatible, extended length
    20 µl
    -
    -
    49,45 mm
    Clear
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 Items
    1 * 960 Items - 333,00 kr
    333,00 kr
    1 * 960 Items
       333,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    613-7414
    Environmentally Preferable
    Miljøcertificering
  • LTS compatible
    200 µl
    -
    -
    50,29 mm
    Clear
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 Items
    1 * 960 Items - 344,00 kr
    344,00 kr
    1 * 960 Items
       344,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    613-7417
    Environmentally Preferable
    Miljøcertificering
  • LTS compatible, extended length
    1000 µl
    -
    -
    98,58 mm
    Clear
    8 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 768 Items
    1 * 768 Items - 333,00 kr
    333,00 kr
    1 * 768 Items
       333,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    613-7411
    Environmentally Preferable
    Miljøcertificering
Tip station stacked rack, non-filtered, sterile
Product Information
  • Spidstype
    Volumen
    Steril
    Filter
    Længde
    Spits farge
    Pakket
    Availability
    Price
  • LTS compatible
    200 µl
    +
    -
    50,29 mm
    Clear
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 Items
    1 * 960 Items - 574,00 kr
    574,00 kr
    1 * 960 Items
       574,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    613-7419
    Environmentally Preferable
    Miljøcertificering
Tip station stacked rack, non-filtered, non sterile
Product Information
  • Spidstype
    Volumen
    Steril
    Filter
    Længde
    Spits farge
    Pakket
    Availability
    Price
  • LTS compatible
    200 µl
    -
    50,29 mm
    Clear
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 Items
    1 * 960 Items - 522,00 kr
    522,00 kr
    1 * 960 Items
       522,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    613-7418
    Environmentally Preferable
    Miljøcertificering
HybridRack, filtered, sterile
Product Information
  • Spidstype
    Volumen
    Steril
    Filter
    Længde
    Spits farge
    Pakket
    Availability
    Price
  • LTS compatible, extended length
    20 µl
    +
    +
    49,45 mm
    Clear
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 Items
    1 * 960 Items - 830,00 kr
    830,00 kr
    1 * 960 Items
       830,00 kr
    Specifications:
      Lav fremstillingspåvirkning|Bæredygtig emballage
    More
    Specifications
    Cat. No.
    613-7424
    Environmentally Preferable
    Lav fremstillingspåvirkning|Bæredygtig emballage
    Miljøcertificering
    Compared to conventional plastic racked pipet tips, reduces plastic per case by 70%.
    Produced in ISO 14001 Certified facility.
      Lav fremstillingspåvirkning|Bæredygtig emballage
  • LTS compatible
    200 µl
    +
    +
    50,29 mm
    Clear
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 Items
    1 * 960 Items - 830,00 kr
    830,00 kr
    1 * 960 Items
       830,00 kr
    Specifications:
      Lav fremstillingspåvirkning|Bæredygtig emballage
    More
    Specifications
    Cat. No.
    613-7425
    Environmentally Preferable
    Lav fremstillingspåvirkning|Bæredygtig emballage
    Miljøcertificering
    Compared to conventional plastic racked pipet tips, reduces plastic per case by 70%.
    Produced in ISO 14001 Certified facility.
      Lav fremstillingspåvirkning|Bæredygtig emballage
  • LTS compatible, extended length
    1000 µl
    +
    +
    98,58 mm
    Clear
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 Items
    1 * 960 Items - 1.070,00 kr
    1.070,00 kr
    1 * 960 Items
       1.070,00 kr
    Specifications:
      Lav fremstillingspåvirkning|Bæredygtig emballage
    More
    Specifications
    Cat. No.
    613-7423
    Environmentally Preferable
    Lav fremstillingspåvirkning|Bæredygtig emballage
    Miljøcertificering
    Compared to conventional plastic racked pipet tips, reduces plastic per case by 70%.
    Produced in ISO 14001 Certified facility.
      Lav fremstillingspåvirkning|Bæredygtig emballage