Quantitative filter papers, ashless grades, MN 640
Om denne vare
These quantitative filter papers are made from refined pulp and linters. They are purified in a highly developed clean-up process to remove organic and inorganic impurities. This ensures a low ash content of <0,01 %. They are consistent performance and give safe results in gravimetric analyses.
- Very low ash content
- Suitable for gravimetric, quantitative analysis
- Contamination-free filtration
Grade MN 640 dd 'green ribbon' is an ashless, slow filter paper which is slightly less retentive than MN 640 d.
Grade MN 640 d 'blue ribbon' is an ashless, slow filter paper with very fine particle retention. Standard paper for critical gravimetric analysis of fine precipitates like barium sulfate or metastannic acid.
Grade MN 640 de is an ashless, very slow filter paper for the filtration of extremely fine particles in quantitative range.
Grade MN 640 m 'white ribbon' is the classic, general purpose, ashless filter paper with medium speed and medium retention. Typical applications include gravimetric analysis for components, quantitative determinations of sediments and high purity clean up procedures.
Grade MN 640 md 'red ribbon' is an ashless filter paper, which is slightly more retentive than MN 640 m with corresponding increase in filtration time.
Grade MN 640 we is the fastest quantitative filter paper with highest flow rate. It is used for example in analytical procedures of samples with coarse or gelatinous particles.
Grade MN 640 w 'black ribbon' is a fast, ashless filter paper. Recommended for analytical procedures involving coarse particles or gelatinous precipitates.
63 tilgængelige muligheder nedenfor
Product Details & Documents
These quantitative filter papers are made from refined pulp and linters. They are purified in a highly developed clean-up process to remove organic and inorganic impurities. This ensures a low ash content of <0,01 %. They are consistent performance and give safe results in gravimetric analyses.
- Very low ash content
- Suitable for gravimetric, quantitative analysis
- Contamination-free filtration
Grade MN 640 dd 'green ribbon' is an ashless, slow filter paper which is slightly less retentive than MN 640 d.
Grade MN 640 d 'blue ribbon' is an ashless, slow filter paper with very fine particle retention. Standard paper for critical gravimetric analysis of fine precipitates like barium sulfate or metastannic acid.
Grade MN 640 de is an ashless, very slow filter paper for the filtration of extremely fine particles in quantitative range.
Grade MN 640 m 'white ribbon' is the classic, general purpose, ashless filter paper with medium speed and medium retention. Typical applications include gravimetric analysis for components, quantitative determinations of sediments and high purity clean up procedures.
Grade MN 640 md 'red ribbon' is an ashless filter paper, which is slightly more retentive than MN 640 m with corresponding increase in filtration time.
Grade MN 640 we is the fastest quantitative filter paper with highest flow rate. It is used for example in analytical procedures of samples with coarse or gelatinous particles.
Grade MN 640 w 'black ribbon' is a fast, ashless filter paper. Recommended for analytical procedures involving coarse particles or gelatinous precipitates.
Specifications for Product Family
Specifikationer
- Catalog No:
- MANA205005
- 512-0495
- 512-0496
- 512-0497
- 515-9800
- MANA205015
- MANA205018
- MANA205024
- MANA205032
- MANA206005
- MANA206007
- MANA206009
- MANA206011
- MANA206012
- MANA206015
- MANA206018
- MANA206024
- MANA206032
- MANA207005
- MANA207007
- MANA207009
- MANA207011
- MANA207012
- MANA207015
- MANA207018
- MANA207024
- MANA207032
- MANA203005
- 512-0490
- MANA203009
- 512-0491
- 515-9710
- 512-0492
- 512-0493
- MANA203024
- MANA203032
- MANA204005
- MANA204007
- MANA204009
- MANA204011
- 512-0494
- MANA204015
- MANA204018
- MANA204024
- MANA204032
- 512-0084
- 512-0486
- 515-9700
- 512-0487
- 515-9701
- 512-0488
- MANA202018
- 512-0489
- MANA202032
- MANA201005
- MANA201007
- MANA201009
- MANA201011
- MANA201012
- MANA201015
- MANA201018
- MANA201024
- MANA201032
- Kvalitet:
- MN 640 d
- MN 640 d
- MN 640 d
- MN 640 d
- MN 640 d
- MN 640 d
- MN 640 d
- MN 640 d
- MN 640 d
- MN 640 dd
- MN 640 dd
- MN 640 dd
- MN 640 dd
- MN 640 dd
- MN 640 dd
- MN 640 dd
- MN 640 dd
- MN 640 dd
- MN 640 de
- MN 640 de
- MN 640 de
- MN 640 de
- MN 640 de
- MN 640 de
- MN 640 de
- MN 640 de
- MN 640 de
- MN 640 m
- MN 640 m
- MN 640 m
- MN 640 m
- MN 640 m
- MN 640 m
- MN 640 m
- MN 640 m
- MN 640 m
- MN 640 md
- MN 640 md
- MN 640 md
- MN 640 md
- MN 640 md
- MN 640 md
- MN 640 md
- MN 640 md
- MN 640 md
- MN 640 w
- MN 640 w
- MN 640 w
- MN 640 w
- MN 640 w
- MN 640 w
- MN 640 w
- MN 640 w
- MN 640 w
- MN 640 we
- MN 640 we
- MN 640 we
- MN 640 we
- MN 640 we
- MN 640 we
- MN 640 we
- MN 640 we
- MN 640 we
- Vægt:
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 100 g/m²
- 100 g/m²
- 100 g/m²
- 100 g/m²
- 100 g/m²
- 100 g/m²
- 100 g/m²
- 100 g/m²
- 100 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- Tykkelse:
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,22 mm
- 0,22 mm
- 0,22 mm
- 0,22 mm
- 0,22 mm
- 0,22 mm
- 0,22 mm
- 0,22 mm
- 0,22 mm
- Partikelretention:
- 2 - 4 µm
- 2 - 4 µm
- 2 - 4 µm
- 2 - 4 µm
- 2 - 4 µm
- 2 - 4 µm
- 2 - 4 µm
- 2 - 4 µm
- 2 - 4 µm
- -
- -
- -
- -
- -
- -
- -
- -
- -
- 1 - 2 µm
- 1 - 2 µm
- 1 - 2 µm
- 1 - 2 µm
- 1 - 2 µm
- 1 - 2 µm
- 1 - 2 µm
- 1 - 2 µm
- 1 - 2 µm
- 4 - 12 µm
- 4 - 12 µm
- 4 - 12 µm
- 4 - 12 µm
- 4 - 12 µm
- 4 - 12 µm
- 4 - 12 µm
- 4 - 12 µm
- 4 - 12 µm
- 4 - 7 µm
- 4 - 7 µm
- 4 - 7 µm
- 4 - 7 µm
- 4 - 7 µm
- 4 - 7 µm
- 4 - 7 µm
- 4 - 7 µm
- 4 - 7 µm
- 7 - 12 µm
- 7 - 12 µm
- 7 - 12 µm
- 7 - 12 µm
- 7 - 12 µm
- 7 - 12 µm
- 7 - 12 µm
- 7 - 12 µm
- 7 - 12 µm
- 12 - 25 µm
- 12 - 25 µm
- 12 - 25 µm
- 12 - 25 µm
- 12 - 25 µm
- 12 - 25 µm
- 12 - 25 µm
- 12 - 25 µm
- 12 - 25 µm
- Filtreringshastighed:
- 140 s (DIN 53137)
- 140 s (DIN 53137)
- 140 s (DIN 53137)
- 140 s (DIN 53137)
- 140 s (DIN 53137)
- 140 s (DIN 53137)
- 140 s (DIN 53137)
- 140 s (DIN 53137)
- 140 s (DIN 53137)
- 100 s (DIN 53137)
- 100 s (DIN 53137)
- 100 s (DIN 53137)
- 100 s (DIN 53137)
- 100 s (DIN 53137)
- 100 s (DIN 53137)
- 100 s (DIN 53137)
- 100 s (DIN 53137)
- 100 s (DIN 53137)
- 195 s (DIN 53137)
- 195 s (DIN 53137)
- 195 s (DIN 53137)
- 195 s (DIN 53137)
- 195 s (DIN 53137)
- 195 s (DIN 53137)
- 195 s (DIN 53137)
- 195 s (DIN 53137)
- 195 s (DIN 53137)
- 27 s (DIN 53137)
- 27 s (DIN 53137)
- 27 s (DIN 53137)
- 27 s (DIN 53137)
- 27 s (DIN 53137)
- 27 s (DIN 53137)
- 27 s (DIN 53137)
- 27 s (DIN 53137)
- 27 s (DIN 53137)
- 55 s (DIN 53137)
- 55 s (DIN 53137)
- 55 s (DIN 53137)
- 55 s (DIN 53137)
- 55 s (DIN 53137)
- 55 s (DIN 53137)
- 55 s (DIN 53137)
- 55 s (DIN 53137)
- 55 s (DIN 53137)
- 9 s (DIN 53137)
- 9 s (DIN 53137)
- 9 s (DIN 53137)
- 9 s (DIN 53137)
- 9 s (DIN 53137)
- 9 s (DIN 53137)
- 9 s (DIN 53137)
- 9 s (DIN 53137)
- 9 s (DIN 53137)
- 5 s (DIN 53137)
- 5 s (DIN 53137)
- 5 s (DIN 53137)
- 5 s (DIN 53137)
- 5 s (DIN 53137)
- 5 s (DIN 53137)
- 5 s (DIN 53137)
- 5 s (DIN 53137)
- 5 s (DIN 53137)
- Askeindhold:
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
- <0,01%
Specifikationer
Product Family Options
Grade MN 640 d, circles
Produktinformation
- KvalitetStørrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.MANA205005KvalitetMN 640 dVægt85 g/m²Tykkelse0,17 mmPartikelretention2 - 4 µmFiltreringshastighed140 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.512-0495KvalitetMN 640 dVægt85 g/m²Tykkelse0,17 mmPartikelretention2 - 4 µmFiltreringshastighed140 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.512-0496KvalitetMN 640 dVægt85 g/m²Tykkelse0,17 mmPartikelretention2 - 4 µmFiltreringshastighed140 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.512-0497KvalitetMN 640 dVægt85 g/m²Tykkelse0,17 mmPartikelretention2 - 4 µmFiltreringshastighed140 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.515-9800KvalitetMN 640 dVægt85 g/m²Tykkelse0,17 mmPartikelretention2 - 4 µmFiltreringshastighed140 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA205015KvalitetMN 640 dVægt85 g/m²Tykkelse0,17 mmPartikelretention2 - 4 µmFiltreringshastighed140 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA205018KvalitetMN 640 dVægt85 g/m²Tykkelse0,17 mmPartikelretention2 - 4 µmFiltreringshastighed140 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA205024KvalitetMN 640 dVægt85 g/m²Tykkelse0,17 mmPartikelretention2 - 4 µmFiltreringshastighed140 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA205032KvalitetMN 640 dVægt85 g/m²Tykkelse0,17 mmPartikelretention2 - 4 µmFiltreringshastighed140 s (DIN 53137)Askeindhold<0,01%
Grade MN 640 dd, circles
Produktinformation
- KvalitetStørrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.MANA206005KvalitetMN 640 ddVægt70 g/m²Tykkelse0,16 mmPartikelretention-Filtreringshastighed100 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA206007KvalitetMN 640 ddVægt70 g/m²Tykkelse0,16 mmPartikelretention-Filtreringshastighed100 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA206009KvalitetMN 640 ddVægt70 g/m²Tykkelse0,16 mmPartikelretention-Filtreringshastighed100 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA206011KvalitetMN 640 ddVægt70 g/m²Tykkelse0,16 mmPartikelretention-Filtreringshastighed100 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA206012KvalitetMN 640 ddVægt70 g/m²Tykkelse0,16 mmPartikelretention-Filtreringshastighed100 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA206015KvalitetMN 640 ddVægt70 g/m²Tykkelse0,16 mmPartikelretention-Filtreringshastighed100 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA206018KvalitetMN 640 ddVægt70 g/m²Tykkelse0,16 mmPartikelretention-Filtreringshastighed100 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA206024KvalitetMN 640 ddVægt70 g/m²Tykkelse0,16 mmPartikelretention-Filtreringshastighed100 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA206032KvalitetMN 640 ddVægt70 g/m²Tykkelse0,16 mmPartikelretention-Filtreringshastighed100 s (DIN 53137)Askeindhold<0,01%
Grade MN 640 de, circles
Produktinformation
- KvalitetStørrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.MANA207005KvalitetMN 640 deVægt100 g/m²Tykkelse0,20 mmPartikelretention1 - 2 µmFiltreringshastighed195 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA207007KvalitetMN 640 deVægt100 g/m²Tykkelse0,20 mmPartikelretention1 - 2 µmFiltreringshastighed195 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA207009KvalitetMN 640 deVægt100 g/m²Tykkelse0,20 mmPartikelretention1 - 2 µmFiltreringshastighed195 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA207011KvalitetMN 640 deVægt100 g/m²Tykkelse0,20 mmPartikelretention1 - 2 µmFiltreringshastighed195 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA207012KvalitetMN 640 deVægt100 g/m²Tykkelse0,20 mmPartikelretention1 - 2 µmFiltreringshastighed195 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA207015KvalitetMN 640 deVægt100 g/m²Tykkelse0,20 mmPartikelretention1 - 2 µmFiltreringshastighed195 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA207018KvalitetMN 640 deVægt100 g/m²Tykkelse0,20 mmPartikelretention1 - 2 µmFiltreringshastighed195 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA207024KvalitetMN 640 deVægt100 g/m²Tykkelse0,20 mmPartikelretention1 - 2 µmFiltreringshastighed195 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA207032KvalitetMN 640 deVægt100 g/m²Tykkelse0,20 mmPartikelretention1 - 2 µmFiltreringshastighed195 s (DIN 53137)Askeindhold<0,01%
Grade MN 640 m, circles
Produktinformation
- KvalitetStørrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.MANA203005KvalitetMN 640 mVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 12 µmFiltreringshastighed27 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.512-0490KvalitetMN 640 mVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 12 µmFiltreringshastighed27 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA203009KvalitetMN 640 mVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 12 µmFiltreringshastighed27 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.512-0491KvalitetMN 640 mVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 12 µmFiltreringshastighed27 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.515-9710KvalitetMN 640 mVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 12 µmFiltreringshastighed27 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.512-0492KvalitetMN 640 mVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 12 µmFiltreringshastighed27 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.512-0493KvalitetMN 640 mVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 12 µmFiltreringshastighed27 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA203024KvalitetMN 640 mVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 12 µmFiltreringshastighed27 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA203032KvalitetMN 640 mVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 12 µmFiltreringshastighed27 s (DIN 53137)Askeindhold<0,01%
Grade MN 640 md, circles
Produktinformation
- KvalitetStørrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.MANA204005KvalitetMN 640 mdVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 7 µmFiltreringshastighed55 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA204007KvalitetMN 640 mdVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 7 µmFiltreringshastighed55 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA204009KvalitetMN 640 mdVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 7 µmFiltreringshastighed55 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA204011KvalitetMN 640 mdVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 7 µmFiltreringshastighed55 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.512-0494KvalitetMN 640 mdVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 7 µmFiltreringshastighed55 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA204015KvalitetMN 640 mdVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 7 µmFiltreringshastighed55 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA204018KvalitetMN 640 mdVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 7 µmFiltreringshastighed55 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA204024KvalitetMN 640 mdVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 7 µmFiltreringshastighed55 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA204032KvalitetMN 640 mdVægt85 g/m²Tykkelse0,20 mmPartikelretention4 - 7 µmFiltreringshastighed55 s (DIN 53137)Askeindhold<0,01%
Grade MN 640 w, circles
Produktinformation
- KvalitetStørrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.512-0084KvalitetMN 640 wVægt85 g/m²Tykkelse0,20 mmPartikelretention7 - 12 µmFiltreringshastighed9 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.512-0486KvalitetMN 640 wVægt85 g/m²Tykkelse0,20 mmPartikelretention7 - 12 µmFiltreringshastighed9 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.515-9700KvalitetMN 640 wVægt85 g/m²Tykkelse0,20 mmPartikelretention7 - 12 µmFiltreringshastighed9 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.512-0487KvalitetMN 640 wVægt85 g/m²Tykkelse0,20 mmPartikelretention7 - 12 µmFiltreringshastighed9 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.515-9701KvalitetMN 640 wVægt85 g/m²Tykkelse0,20 mmPartikelretention7 - 12 µmFiltreringshastighed9 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.512-0488KvalitetMN 640 wVægt85 g/m²Tykkelse0,20 mmPartikelretention7 - 12 µmFiltreringshastighed9 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA202018KvalitetMN 640 wVægt85 g/m²Tykkelse0,20 mmPartikelretention7 - 12 µmFiltreringshastighed9 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.512-0489KvalitetMN 640 wVægt85 g/m²Tykkelse0,20 mmPartikelretention7 - 12 µmFiltreringshastighed9 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA202032KvalitetMN 640 wVægt85 g/m²Tykkelse0,20 mmPartikelretention7 - 12 µmFiltreringshastighed9 s (DIN 53137)Askeindhold<0,01%
Grade MN 640 we, circles
Produktinformation
- KvalitetStørrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.MANA201005KvalitetMN 640 weVægt85 g/m²Tykkelse0,22 mmPartikelretention12 - 25 µmFiltreringshastighed5 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA201007KvalitetMN 640 weVægt85 g/m²Tykkelse0,22 mmPartikelretention12 - 25 µmFiltreringshastighed5 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA201009KvalitetMN 640 weVægt85 g/m²Tykkelse0,22 mmPartikelretention12 - 25 µmFiltreringshastighed5 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA201011KvalitetMN 640 weVægt85 g/m²Tykkelse0,22 mmPartikelretention12 - 25 µmFiltreringshastighed5 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA201012KvalitetMN 640 weVægt85 g/m²Tykkelse0,22 mmPartikelretention12 - 25 µmFiltreringshastighed5 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA201015KvalitetMN 640 weVægt85 g/m²Tykkelse0,22 mmPartikelretention12 - 25 µmFiltreringshastighed5 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA201018KvalitetMN 640 weVægt85 g/m²Tykkelse0,22 mmPartikelretention12 - 25 µmFiltreringshastighed5 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA201024KvalitetMN 640 weVægt85 g/m²Tykkelse0,22 mmPartikelretention12 - 25 µmFiltreringshastighed5 s (DIN 53137)Askeindhold<0,01% - Specifikationer:
More
Specifications
Cat. No.MANA201032KvalitetMN 640 weVægt85 g/m²Tykkelse0,22 mmPartikelretention12 - 25 µmFiltreringshastighed5 s (DIN 53137)Askeindhold<0,01%