Cimarec™ VARIOMAG® i Telemodul Controller
Telemodul stirrer controller, 230 V, operating at 0...40 °C (80% relative humidity), 96×63×50 mm
Katalog # HPLT50119118
Leverandør: THERMO H + P LABORTECHNIK
Cimarec™ VARIOMAG® i Telemodul Controller
Katalog # HPLT50119118
Leverandør: THERMO H + P LABORTECHNIK
Gå til:
Product Details & Documents
Save space with Thermo Scientific™ Cimarec™ i Telemodul controllers, including Micro, Mini, Compact and Maxi stirrers.
- Provide stirring power for small to medium volumes of low-viscosity fluids
- Recommended for single point stirring up to 250 ml
Ordering information: Order as replacement controller only.
Specifikationer
- Beskrivelse:Telemodul stirrer controller, 230 V, operating at 0...40 °C (80% relative humidity), 96×63×50 mm