Orders LinkContactUsLinkComponent
STEMscript™ cDNA Synthesis Kit with Oligo(dT) Primers
STEMscript™ cDNA Synthesis Kit with Oligo(dT) Primers
STEMscript™ cDNA synthesis kit with oligo(dT) primers, 1 KIT
Katalog # STMC79003
Marketsource
Leverandør:   Stemcell Technologies
STEMscript™ cDNA Synthesis Kit with Oligo(dT) Primers
Katalog # STMC79003
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
STEMscript™ cDNA Synthesis Kit with Oligo(dT) Primers
STEMscript™ cDNA Synthesis Kit with Oligo(dT) Primers
STEMscript™ cDNA synthesis kit with oligo(dT) primers, 1 KIT
CAS nummer:  
Price
4.121,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
For reverse transcription of eukaryotic mRNA before qPCR or RNA sequencing.
Use STEMscript™ cDNA Synthesis Kit with Oligo(dT) Primers for reverse transcription of eukaryotic mRNA prior to qPCR or RNA sequencing. This kit includes three components that have been optimised to provide efficient synthesis of first-strand cDNA in preparation for qPCR amplification: STEMscript™ Reverse Transcriptase Mix (contains STEMscript™ Reverse Transcriptase purified from recombinant E. coli, and recombinant ribonuclease inhibitor), STEMscript™ Reaction Buffer (contains primers, MgCl₂, and dNTPs), and nuclease-free water.
Oligo(dT) primers are ideal for reverse transcription of eukaryotic mRNA, as they anneal to poly(A) tails. For reverse transcription of RNA without poly(A) tails (e.g. rRNA, tRNA, degraded RNA, RNA isolated from prokaryotic sources, and RNA with known secondary structures), we recommend using STEMscript™ cDNA Synthesis Kit with Random Primers.
Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    STEMscript™ cDNA synthesis kit with oligo(dT) primers