Orders LinkContactUsLinkComponent
ArciTect™ Human HPRT Positive Control Kit
ArciTect™ Human HPRT Positive Control Kit
ArciTect™ Human HPRT positive control kit, 1 KIT
Katalog # STMC76013
Marketsource
Leverandør:   Stemcell Technologies
ArciTect™ Human HPRT Positive Control Kit
Katalog # STMC76013
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
ArciTect™ Human HPRT Positive Control Kit
ArciTect™ Human HPRT Positive Control Kit
ArciTect™ Human HPRT positive control kit, 1 KIT
CAS nummer:  
Price
2.867,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Positive control for CRISPR-Cas9 genome editing.
ArciTect™ Human HPRT Positive Control Kit is designed as a positive control for experiments using the ArciTect™ CRISPR-Cas9 genome editing system. The kit comprises ArciTect™ Human HPRT crRNA (2 nmol), ArciTect™ Human HPRT Primer Mix (2 nmol), ArciTect™ tracrRNA (5 nmol), and ArciTect™ Annealing Buffer (5X). ArciTect™ Human HPRT crRNA first requires annealing to ArciTect™ tracrRNA (included in the kit). This must then be combined with an ArciTect™ Cas9 Nuclease to form a ribonucleoprotein complex. ArciTect™ Human HPRT Primer Mix can be used to amplify genomic DNA isolated from a population of transfected cells, which can subsequently be used in a T7 endonuclease I assay to determine cleavage efficiency.
This kit has been tested and validated for use with the ArciTect™ line of genome editing products. HPRT, or hypoxanthine phosphoribosyltransferase, is a housekeeping gene and a commonly used control. The kit can be used to optimize transfection protocols and act as a positive control that can be used alongside custom ArciTect™ crRNAs (e.g. Cat. #STMC76010).
Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    ArciTect™ Human HPRT positive control kit
  • Cat. No.:
    STMC76013
  • Cellespecifik:
    Pluripotent stem cells
Specifikationer