Orders LinkContactUsLinkComponent
SpinSep™ Density Medium
SpinSep™ Density Medium
SpinSep™ density medium
Katalog # STMC17531
Marketsource
Leverandør:   Stemcell Technologies
SpinSep™ Density Medium
Katalog # STMC17531
Leverandør:  Stemcell Technologies
Marketsource
SpinSep™ Density Medium
SpinSep™ Density Medium
SpinSep™ density medium
Price
1.470,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
Density gradient medium.
  • For use in cell isolation applications
SpinSep™ Density medium is a density gradient medium designed for use with the EasySep™ human myeloid positive selection kit.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifikationer
  • Beskrivelse:
    SpinSep™ density medium