Solid Glass Stopper
Solid glass stopper, clear, 34/35
Katalog # NEUB1641-91-3435
Leverandør: NEUBERT VOLUME GLASSWAERE
Gå til:
Product Details & Documents
Six sided, solid glass stopper.
- Made from Borosilicate glass 3.3
- Six point handle
Specifikationer
- NS:34/35
- Beskrivelse:Solid glass stopper, clear