1,2-Dibromethan-D4 98% til NMR spektroskopi, for environmental analysis
Supplier: CAMBRIDGE ISOTOPE LABS
New Product
Certificates
About this item
CAS Number:22581-63-1
:
Formula:BrH₂CCH₂Br
MDL Number:MFCD00037499
Boiling Pt:131,..132 °C (lit.)
Storage Temperature:Room Temperature
EINECS:245-102-8
UN:1605
ADR:6.1,I
:191.83 g/mol
:GHS06
:GHS08
:GHS09
Frequently Bought Together