Orders LinkContactUsLinkComponent
PCA - Plate Count Agar
PCA - Plate Count Agar
Plate count agar (PCA), 9×250 ml/bottle, Glass bottle, 9
Katalog # DOEH876631782
Leverandør:   DOEHLER GROUP
PCA - Plate Count Agar
Katalog # DOEH876631782
Leverandør:  DOEHLER GROUP
CAS nummer:  
PCA - Plate Count Agar
PCA - Plate Count Agar
Plate count agar (PCA), 9×250 ml/bottle, Glass bottle, 9
CAS nummer:  
Price
681,00 kr
Log ind for tilgængelighed
1 * 9 Items
Product Details & Documents
Agar medium for the quantitative detection of the total germ count in the beverage and food industry.
  • Ready-to-use, liquefaction possible directly in the bottle
  • Universal applicable (membrane filtration, pouring and spread plate method)
  • Reference medium, pH neutral
The number of detectable microorganisms is an important indicator for quality assurance. The medium is also known as Bacteria Count Agar or Caseine-peptone Dextrose Yeast Agar and complies with the requirements of EN ISO 4833 and ISO 17410.
Documents
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle
  • Pakn:
    9
  • Beskrivelse:
    Plate count agar (PCA)
  • Størrelse:
    9×250 ml/bottle