Orders LinkContactUsLinkComponent
Inductively Coupled Plasma-Mass Spectrometry (ICP-MS)
Inductively Coupled Plasma-Mass Spectrometry (ICP-MS)
Katalog #
Inductively Coupled Plasma-Mass Spectrometry (ICP-MS)
Katalog #
Leverandør:  
Inductively Coupled Plasma-Mass Spectrometry (ICP-MS)
Inductively Coupled Plasma-Mass Spectrometry (ICP-MS)
Product Details & Documents
Agilent ICP-MS systems offer unmatched sensitivity, accuracy, and interference removal for trace elemental analysis across a wide range of sample types. With both single and triple quadrupole configurations, Agilent ICP-MS instruments deliver high matrix tolerance, broad dynamic range, and robust performance even in the most challenging applications. Whether you're conducting routine environmental testing, pharmaceutical QA/QC, or advanced research, Agilent ICP-MS solutions provide the confidence, speed, and precision your lab demands.
  • Accurate results at all concentrations, even in complex matrices
  • Reduced sample prep with Ultra High Matrix Introduction (UHMI) technology analyze samples with up to 25% total dissolved solids
  • Minimized downtime through Early Maintenance Feedback (EMF) with proactive alerts
  • Simplified method development with predefined methods and SOPs for routine and regulated analyses
  • Confident interference removal using helium collision cell and MS/MS technology (8900 ICP-QQQ)
  • High throughput and productivity, ideal for routine labs and advanced research
Single and triple quadrupole options: 7850 (SQ), 7900 (advanced SQ), and 8900 (Triple Quad). Helium collision cell and half-mass correction: Resolves polyatomic and doubly charged interferences. Orthogonal detector system: Delivers high sensitivity and wide dynamic range (up to 11 orders of magnitude). Fast transient signal detection: Supports laser ablation, nanoparticle, and chromatographic analyses. MS/MS Reaction cell (8900): Enables precise control of reaction chemistry for complex samples. Color-coded maintenance alerts: Ensures timely and efficient instrument upkeep.
Matrix tolerance: Analyze samples with up to 25% total dissolved solids using Ultra High Matrix Introduction (UHMI) technology
Dynamic range: Wide dynamic range-up to 11 orders of magnitude with the orthogonal detector system
Interference removal:
Helium collision cell and half-mass correction (7850, 7900)
MS/MS reaction cell for advanced interference control (8900 ICP-QQQ)
Detection capabilities: High sensitivity for trace-level detection across a broad concentration range
Transient signal detection: Supports laser ablation, nanoparticle, and chromatographic analyses
Maintenance: Early Maintenance Feedback (EMF) with color-coded alerts for proactive servicing
Software integration: Predefined methods, SOPs, and LIMS compatibility for streamlined workflows
Application fit:
7850: Routine and regulated labs
7900: High matrix and advanced research
8900: Semiconductor, speciation, and ultra-trace analysis
Ordering information: Each model is configurable for your needs. Contact your Avantor representative for more information.