Supelco® Organic Reference Standard, Ethyl 4-Hydroxybenzoate-Ring-¹³C₆ Solution
Ethyl 4-hydroxybenzoate-ring-¹³C₆ solution, Analytical standard, 1 mL
Katalog # 32559-1ML
Leverandør: Merck
Supelco® Organic Reference Standard, Ethyl 4-Hydroxybenzoate-Ring-¹³C₆ Solution
Katalog # 32559-1ML
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
- GHS signal: GHS02, GHS07
Advarsel: P210 - P233 - P240 - P241 - P242 - P305 + P351 + P338
Specifikationer
Specifikationer
- Pakn:1 mL
- Standard:Analytical standard
- Beskrivelse:Ethyl 4-hydroxybenzoate-ring-¹³C₆ solution
- Cat. No.:32559-1ML
- M-Clarity quality level:MQ100
- Molekylvægt:172,13
- Fareklassebeskrivelse:H225 - H319 - H336
- Opbevaringstemperatur:−20 °C
- Flammepunkt:–17,0 °C - closed cup
- Opbevaring betingelser:–20 °C
- Synonym(s):4-Hydroxybenzoic acid ethyl ester-ring-¹³C₆ solution; Ethyl paraben-ring-¹³C₆ solution
- Shipping conditions:WICE
- Formel:13C₆C₃H₁₀O₃
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



