Orders LinkContactUsLinkComponent
Live or Dead™ Fixable Dead Cell Staining Kit
Live or Dead™ Fixable Dead Cell Staining Kit
200 Tests
Katalog # AATB22598
Leverandør:   AAT Bioquest
Live or Dead™ Fixable Dead Cell Staining Kit
Katalog # AATB22598
Leverandør:  AAT Bioquest
CAS nummer:  
Live or Dead™ Fixable Dead Cell Staining Kit
200 Tests
CAS nummer:  
Price
2.010,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
IR Fluorescence. Our Live or Dead™ fixable dead cell staining kits are a set of tools for labelling cells for fluorescence microscopic investigations of cellular functions.
  • The kit provides all the essential components with an optimized cell-labelling protocol
  • The fluorescent dye used in the kit is well excited with the IR laser (808 nm) and emits fluorescence at 878 nm (885/40 bandpass filter).
This particular kit is designed to uniformly label fixed mammalian cells in infrared fluorescence for flow cytometry applications with IR laser excitation.
Leveringsinformation: Shipped at RT.
Advarsel: Research use only.
Specifikationer
Specifikationer
  • Pakn:
    200 Tests