Amplite® Rapid Colorimetric APC-Maleimide Quantitation Kit
Catalog # AATB5527
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- Pakn:2 Tests
- Test pr. sæt:2 tests
Specifications
About this item
Designed for consistent and dependable results. The Amplite® Rapid Colorimetric APC-Maleimide Quantitation Kit facilitates the quantification of maleimide groups on maleimide-activated APC proteins using a proprietary sensor, Maleimide 555™, which has an absorption peak at ~555 nm.
- Robust protocol
- This quantitation kit is compatible with various detection systems, including traditional cuvettes, NanoDrop™ Spectrophotometers, and 96-well absorbance plate readers
- Six-month shelf life
Maleimide-based crosslinking reagents are commonly used for conjugating proteins to other proteins or biomolecules.
Delivery information: Shipped with blue ice.
Caution: Research use only.