Orders LinkContactUsLinkComponent
Cell Meter™ Glucose Uptake Imaging Kit
100 tests, 100 Tests
Katalog # AATB23501
Leverandør:   AAT Bioquest
Cell Meter™ Glucose Uptake Imaging Kit
Katalog # AATB23501
Leverandør:  AAT Bioquest
CAS nummer:  
Cell Meter™ Glucose Uptake Imaging Kit
100 tests, 100 Tests
CAS nummer:  
Price
6.850,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Red Fluorescence. The Cell Meter™ Glucose Uptake Imaging Kit provides a simple and direct method for quantifying glucose uptake across diverse cellular contexts.
  • Ex/Em = 651/670 nm
  • Quantification via both fluorescence microscopy and flow cytometry techniques
  • Six-month shelf life
This kit employs Glutite™ Red 670, a cell-permeable fluorescent glucose tracer, enabling precise molecular sensing and bioimaging via GLUT transporters.
Delivery information: Shipped at RT.
Caution: Research use only.
Specifikationer
Specifikationer
  • Pakn:
    100 Tests
  • Test pr. sæt:
    100 tests
Specifikationer