Orders LinkContactUsLinkComponent
Amplite® Fluorimetric Sodium Ion Quantification Kit
Amplite® Fluorimetric Sodium Ion Quantification Kit
100 tests, 100 Tests
Katalog # AATB21325
Leverandør:   AAT Bioquest
Amplite® Fluorimetric Sodium Ion Quantification Kit
Katalog # AATB21325
Leverandør:  AAT Bioquest
CAS nummer:  
Amplite® Fluorimetric Sodium Ion Quantification Kit
100 tests, 100 Tests
CAS nummer:  
Price
5.220,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Designed for consistent and dependable results. Amplite® fluorimetric sodium ion quantification kit uses our robust sodium ion indicator dye, SoNa™ 520, which exhibit great fluorescence intensity enhancement upon binding to sodium ions.
  • Robust protocol
  • SoNa™ 520 is perhaps the most robust sodium ion indicator with high selectivity
  • 12 month shelf life
This microplate-based assay kit requires extremely small amount of sample, it is particularly suitable for the determination of sodium ion concentration in microvolume format.
Leveringsinformation: Shipped at RT.
Advarsel: Research use only.
Specifikationer
Specifikationer
  • Pakn:
    100 Tests
  • Test pr. sæt:
    100 tests