Wheat Germ Agglutinin, XFD350 Labelled *XFD350 Same Structure to Alexa Fluor® 350*
Catalog # AATB25495
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- Pakn:1 mg
- Beskrivelse:Wheat germ agglutinin, XFD350 labelled *XFD350 same structure to Alexa Fluor® 350*
Specifications
About this item
XFD350, manufactured by AAT Bioquest, has an identical chemical structure to Alexa Fluor® 350, a trademark of ThermoFisher.
- Stable for 12 months
Quality tested in-house.
Caution: For research use only.