Supelco® TraceCERT® Organic Reference Standard, Phosphamidon
Phosphamidon, Certified reference material, TraceCERT®, 100 mg
Katalog # CRM44794-100MG
Leverandør: Merck
Supelco® TraceCERT® Organic Reference Standard, Phosphamidon
Katalog # CRM44794-100MG
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Advarsel: P202 - P264 - P273 - P280 - P301 + P310 - P302 + P352 + P312
Specifikationer
Specifikationer
- Pakn:100 mg
- Standard:Certified reference material, TraceCERT®
- Beskrivelse:Phosphamidon
- Cat. No.:CRM44794-100MG
- M-Clarity quality level:MQ300
- CAS-nr:13171-21-6
- Molekylvægt:299,69
- MDL No.:MFCD00055301
- Fareklassebeskrivelse:H300 - H311 - H341 - H410
- Opbevaring betingelser:–20 °C
- Synonym(s):2-Chloro-N,N-diethyl-3-(dimethylphosphono)crotonic amide
- Shipping conditions:WICE



