Supelco® TraceCERT® Organic Reference Standard, Lithium bromide solution
Lithium bromide solution, Certified reference material, TraceCERT®, 2,5 L
Katalog # 93687-2.5L
Leverandør: Merck
Supelco® TraceCERT® Organic Reference Standard, Lithium bromide solution
Katalog # 93687-2.5L
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Specifikationer
Specifikationer
- Pakn:2,5 L
- Standard:Certified reference material, TraceCERT®
- Beskrivelse:Lithium bromide solution
- Cat. No.:93687-2.5L
- M-Clarity quality level:MQ300
- CAS-nr:7550-35-8
- Molekylvægt:86,85
- MDL No.:MFCD00011077
- Opbevaring betingelser:10...30 °C
- Shipping conditions:STD
- Formel:LiBr



