Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, Temephos

Supelco® TraceCERT® Organic Reference Standard, Temephos

Temephos, Certified reference material, TraceCERT®, 50 mg
Katalog # 75817-50MG
Leverandør:   Merck
Supelco® TraceCERT® Organic Reference Standard, Temephos
Katalog # 75817-50MG
Leverandør:  Merck
CAS nummer:  
Supelco® TraceCERT® Organic Reference Standard, Temephos

Temephos, Certified reference material, TraceCERT®, 50 mg

CAS nummer:  
Price
611,00 kr
Log ind for tilgængelighed
1 * 50 mg
Product Details & Documents

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.

Advarsel: P264 - P270 - P273 - P280 - P301 + P312 - P302 + P352 + P312

Specifikationer

Specifikationer

  • Pakn:
    50 mg
  • Standard:
    Certified reference material, TraceCERT®
  • Beskrivelse:
    Temephos
  • Cat. No.:
    75817-50MG
  • M-Clarity quality level:
    MQ300
  • CAS-nr:
    3383-96-8
  • Molekylvægt:
    466,47
  • MDL No.:
    MFCD00128034
  • Fareklassebeskrivelse:
    H302 - H311 - H410
  • Opbevaring betingelser:
    –20 °C
  • Shipping conditions:
    WICE

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.