Supelco® TraceCERT® Organic Reference Standard, Permethrin
Permethrin, Certified reference material, TraceCERT®, mixture of cis and trans isomers, 100 mg
Katalog # 68216-100MG
Leverandør: Merck
Supelco® TraceCERT® Organic Reference Standard, Permethrin
Katalog # 68216-100MG
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Advarsel: P261 - P273 - P280 - P301 + P312 - P302 + P352 - P304 + P340 + P312
Specifikationer
Specifikationer
- Pakn:100 mg
- Standard:Certified reference material, TraceCERT®, mixture of cis and trans isomers
- Beskrivelse:Permethrin
- Cat. No.:68216-100MG
- M-Clarity quality level:MQ300
- CAS-nr:52645-53-1
- Molekylvægt:391,29
- MDL No.:MFCD00041809
- Fareklassebeskrivelse:H302 + H332 - H317 - H410
- Opbevaring betingelser:–20 °C
- Shipping conditions:WICE
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



