Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

XFD532 tyramide

200
Katalog # AATB11072
Leverandør:   AAT Bioquest
XFD532 tyramide
Katalog # AATB11072
Leverandør:  AAT Bioquest
CAS nummer:  
XFD532 tyramide

200

CAS nummer:  
Price
2.160,00 kr
Log ind for tilgængelighed
1 * 200 Items
Product Details & Documents

XFD 532 tyramide contains the Alexa Fluor® 532 fluorophore that can be readily detected with the Alexa Fluor® 532/ATTO 532 filter set (Alexa Fluor® is the trademark of ThermoFisher).

  • Stable for 12 months

Tyramide signal amplification (TSA) has proven to be a particularly versatile and powerful enzyme amplification technique with improved assay sensitivity.

 : For Research Use Only

  • Molecular Weight:847.06 g/mol
Specifikationer

Specifikationer

  • Pakn:
    200

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.