Orders LinkContactUsLinkComponent
SDA alkyne
SDA alkyne
1 mg
Katalog # AATB39005
Leverandør:   AAT Bioquest
SDA alkyne
Katalog # AATB39005
Leverandør:  AAT Bioquest
CAS nummer:  
SDA alkyne
1 mg
CAS nummer:  
Price
1.000,00 kr
Log ind for tilgængelighed
1 * 1 mg
Product Details & Documents
SDA alkyne contains both diazirine photoaffinity moiety and alkyne functional group.
  • Stable for 24 months
Quality tested in-house
 : For Research Use Only
Specifikationer
Specifikationer
  • Pakn:
    1 mg
Specifikationer