0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
APC-Cy7 maleimide
APC-Cy7 maleimide, ReadiUse™, preactivated
APC-Cy7 maleimide, ReadiUse™, preactivated
Catalog # AATB2722
Supplier:
AAT Bioquest
New Product
CAS Number:
Expand image
APC-Cy7 maleimide, ReadiUse™, preactivated
Catalog # AATB2722
Supplier:
AAT Bioquest
New Product
CAS Number:
Specifications
Pakn:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
Product Details