Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Cy3B alkyne

1 mg
Katalog # AATB944
Leverandør:   AAT Bioquest
Cy3B alkyne
Katalog # AATB944
Leverandør:  AAT Bioquest
CAS nummer:  
Cy3B alkyne

1 mg

CAS nummer:  
Price
4.350,00 kr
Log ind for tilgængelighed
1 * 1 mg
Product Details & Documents

Cy3® dye is one of the most common cyanine dyes used for labeling proteins, nucleic acids and other biological molecules.

  • Stable for 12 months

Quality tested in-house

 : For Research Use Only

  • Molecular Weight:950.15 g/mol
Specifikationer

Specifikationer

  • Pakn:
    1 mg

Specifikationer