Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ac-Leu-AMC

Ac-Leu-AMC

1 mg
Katalog # AATB13444
Leverandør:   AAT Bioquest
Ac-Leu-AMC
Katalog # AATB13444
Leverandør:  AAT Bioquest
CAS nummer:  
Ac-Leu-AMC

1 mg

CAS nummer:  
Price
1.170,00 kr
Log ind for tilgængelighed
1 * 1 mg
Product Details & Documents

Ac-Leu-AMC is a fluorogenic substrate for leucine aminopeptidase.

  • Stable for 24 months

Quality tested in-house

Advarsel: For Research Use Only

  • Molecular Weight:330.38 g/mol
Specifikationer

Specifikationer

  • Pakn:
    1 mg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.