Orders LinkContactUsLinkComponent
ReadiUse™ Yeast Cell Lysis Buffer, 5X
ReadiUse™ Yeast Cell Lysis Buffer, 5X
ReadiUse™ Yeast cell lysis buffer 5X, 10 mL
Katalog # AATB24102
Leverandør:   AAT Bioquest
ReadiUse™ Yeast Cell Lysis Buffer, 5X
Katalog # AATB24102
Leverandør:  AAT Bioquest
CAS nummer:  
ReadiUse™ Yeast Cell Lysis Buffer, 5X
ReadiUse™ Yeast Cell Lysis Buffer, 5X
ReadiUse™ Yeast cell lysis buffer 5X, 10 mL
CAS nummer:  
Price
1.170,00 kr
Log ind for tilgængelighed
1 * 10 mL
Product Details & Documents
ReadiUse™ Yeast Cell Lysis Buffer is a biochemical reagent designed for efficient yeast cell lysis and protein solubilisation in laboratory applications.
  • Designed for ease of use
  • Saves time
  • Versatile in its efficacy across various organisms, including Bacillus subtilis, Gram-positive, and Gram-negative bacteria
With a mild detergent formulation, ReadiUse™ Yeast Cell Lysis Buffer minimises the reliance on harsh chemical treatments, extreme pH adjustments, or temperature variations, contributing to the preservation of extracted protein integrity.
Advarsel: For research use only.
Specifikationer
Specifikationer
  • Pakn:
    10 mL
  • Beskrivelse:
    ReadiUse™ Yeast cell lysis buffer 5X