Orders LinkContactUsLinkComponent
Intelsius® Pharmachill Reusable Cold Pack
Intelsius® Pharmachill Reusable Cold Pack
300×200×30 mm, 1600 g
Katalog # DGINCP023
Leverandør:   DGP INTELSIUS
Intelsius® Pharmachill Reusable Cold Pack
Katalog # DGINCP023
Leverandør:  DGP INTELSIUS
Intelsius® Pharmachill Reusable Cold Pack
300×200×30 mm, 1600 g
Price
504,00 kr
Log ind for tilgængelighed
1 * 8 Items
Product Details & Documents
Pharmachill cold packs are reusable thermal media solutions using a water-based PCM ideal for maintaining payload temeprature at between 2 to 8 ˚C and 15 to 25 ˚C.
  • Reusable
  • Durable hard plastic
  • Phase change material
Intelsius® produces a range of thermal media tailored to varied performance requirements. The Pharmachill range includes rigid, liquid-filled coolant bottles designed for use with temperature controlled packaging solutions.
Pharmachill Cold Packs feature a non-toxic, water based PCM. They have been specifically designed to be reusable, and therefore deliver a cost and environmental saving with each reuse.
Specifikationer
  • B×D×H:
    300×200×30 mm
  • Vægt:
    1600 g