Human ECM29 ELISA Kit
Human ECM29 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ECM29 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Detection range: 15,625 to 1000 pg/ml
- Sensitivity: 9,375 pg/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay precision: Intra - assay: CV <8%, Inter - assay: CV <10%
Human ECM29 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human ECM29 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for ECM29 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the ECM29 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-ECM29 antibody, which binds the captured ECM29 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of ECM29 captured in each well. The concentration of ECM29 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
- Pakn:96 Tests
- Beskrivelse:Human ECM29 ELISA kit
- Assay princip:Quantitative
- Sensitivitet:9,375 pg/ml
- Assay varighed:Multiple steps
- Primary antibody reactivity:Human
- Detektionsområde:15,625 - 1000 pg/ml
- Shelf life:Store for 6 months at 4 °C
- Prøvetype:Serum, plasma, tissue homogenates, and other biological fluids
- Time to Results:4 h 30 min
- Analysetype:Sandwich
- Cat. No.:ANTIA302915-96
- Format:Pre-coated
- Krydsreaktivitet:Human ECM29 ELISA Kit exhibits high specificity and excellent specificity for the detection of human ECM29. No significant cross-reactivity or interference between ECM29 and analogues was observed.
- Target protein:ECM29
- Detektionsmetode:Colorimetric
- Prøvevolumen:100 μl
- Opbevaringstemperatur:4 °C
- Regulatory status:RUO
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



