Biotin ELISA Kit
Product Details & Documents
Biotin ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Biotin in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Biotin ELISA kit employs the competitive enzyme immunoassay technique for the quantitative measurement of universal Biotin in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Biotin antigen. During the incubation, Biotin present in the samples or standards competes with the fixed amount of immobilised Biotin for binding sites on the Biotinylated anti-Biotin antibody. The more Biotin present in a sample or standard, the less Biotinylated anti-Biotin antibody that binds to the plate. Following incubation, unbound Biotinylated anti-Biotin antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Biotin present in each sample or standard. The concentration of Biotin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Specifikationer
Specifikationer
- Pakn:96 Tests
- Beskrivelse:Biotin ELISA kit
- Assay princip:Quantitative
- Sensitivitet:10 pg/ml
- Assay varighed:Multiple steps
- Primary antibody reactivity:Other
- Detektionsområde:100 - 2000 pg/ml
- Shelf life:Store for 6 months at 4 °C
- Prøvetype:Serum, plasma, tissue homogenates, and other biological fluids
- Time to Results:4 h 30 min
- Analysetype:Competitive
- Cat. No.:ANTIA247122-96
- Format:Pre-coated
- Krydsreaktivitet:Biotin ELISA Kit exhibits high specificity and excellent specificity for the detection of universal Biotin. No significant cross-reactivity or interference between Biotin and analogues was observed.
- Target protein:Biotin
- Detektionsmetode:Colorimetric
- Prøvevolumen:50 μl
- Opbevaringstemperatur:4 °C
- Regulatory status:RUO



