Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human ZIP-4 ELISA Kit

Human ZIP-4 ELISA Kit

Human ZIP-4 ELISA kit, 96 T
Katalog # ANTIA247062-96
Leverandør:   Antibodies.com
Human ZIP-4 ELISA Kit
Katalog # ANTIA247062-96
Leverandør:  Antibodies.com
CAS nummer:  
Human ZIP-4 ELISA Kit
Human ZIP-4 ELISA Kit

Human ZIP-4 ELISA kit, 96 T

CAS nummer:  
Price
4.670,00 kr
Log ind for tilgængelighed
1 * 96 T
Product Details & Documents

Human ZIP-4 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ZIP-4 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human ZIP-4 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human ZIP-4 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for ZIP-4 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the ZIP-4 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-ZIP-4 Antibody, which binds the captured ZIP-4 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of ZIP-4 captured in each well. The concentration of ZIP-4 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Bestillingsoplysninger: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifikationer

Specifikationer

  • Pakn:
    96 T
  • Beskrivelse:
    Human ZIP-4 ELISA kit
  • Sensitivitet:
    0,094 ng/ml
  • Assay varighed:
    Multiple steps
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    0,156 - 10 ng/ml
  • Shelf life:
    Store for 6 months at 4 °C
  • Prøvetype:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Time to Results:
    4 h 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ANTIA247062-96
  • Format:
    Pre-coated
  • Krydsreaktivitet:
    Human ZIP-4 ELISA Kit exhibits high specificity and excellent specificity for the detection of human ZIP-4. No significant cross-reactivity or interference between ZIP-4 and analogues was observed.
  • Target protein:
    ZIP-4
  • Detektionsmetode:
    Colorimetric
  • Prøvevolumen:
    100 μl
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO

Specifikationer