Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mouse iNOS ELISA Kit

Mouse iNOS ELISA Kit

Mouse iNOS ELISA kit, 96 Tests
Katalog # ANTIA74102-96
Leverandør:   Antibodies.com
Mouse iNOS ELISA Kit
Katalog # ANTIA74102-96
Leverandør:  Antibodies.com
CAS nummer:  
Mouse iNOS ELISA Kit
Mouse iNOS ELISA Kit

Mouse iNOS ELISA kit, 96 Tests

CAS nummer:  
Price
4.670,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Mouse iNOS ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse iNOS in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Detection Range: 0,313 to 20 ng/ml
  • Sensitivity: 0,188 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Mouse iNOS ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse iNOS in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for iNOS has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the iNOS present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-iNOS Antibody, which binds the captured iNOS present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of iNOS captured in each well. The concentration of iNOS can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Bestillingsoplysninger: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifikationer

Specifikationer

  • Pakn:
    96 Tests
  • Beskrivelse:
    Mouse iNOS ELISA kit
  • Assay princip:
    Quantitative
  • Sensitivitet:
    0,188 ng/ml
  • Assay varighed:
    Multiple steps
  • Primary antibody reactivity:
    Mouse
  • Detektionsområde:
    0,313 - 20 ng/ml
  • Shelf life:
    Store for 6 months at 4 °C
  • Prøvetype:
    Serum, plasma, tissue homogenates and other biological fluids
  • Time to Results:
    4 h 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ANTIA74102-96
  • Format:
    Pre-coated
  • Krydsreaktivitet:
    Mouse iNOS ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse iNOS. No significant cross-reactivity or interference between iNOS and analogues was observed.
  • Target protein:
    iNOS
  • Detektionsmetode:
    Colorimetric
  • Prøvevolumen:
    100 μl
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.