Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human NADH2 ELISA Kit

Human NADH2 ELISA Kit

Human NADH2 ELISA kit, 96 Tests
Katalog # ANTIA313283-96
Leverandør:   Antibodies.com
Human NADH2 ELISA Kit
Katalog # ANTIA313283-96
Leverandør:  Antibodies.com
CAS nummer:  
Human NADH2 ELISA Kit
Human NADH2 ELISA Kit

Human NADH2 ELISA kit, 96 Tests

CAS nummer:  
Price
4.220,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Human NADH2 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human NADH2 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 150 to 2400 ng/L
  • Sensitivity: 9,04 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human NADH2 ELISA kit (A313283) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human NADH2 in serum, plasma or other biological fluids. An antibody specific for NADH2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the NADH2 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-NADH2 antibody, which also binds the NADH2 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-NADH2 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-NADH2 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of NADH2 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of NADH2 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Bestillingsoplysninger: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Specifikationer

Specifikationer

  • Pakn:
    96 Tests
  • Beskrivelse:
    Human NADH2 ELISA kit
  • Sensitivitet:
    9,04 ng/L
  • Assay varighed:
    One step
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    150 - 2400 ng/L
  • Shelf life:
    Store for 6 months at 4 °C
  • Prøvetype:
    Serum, plasma or other biological fluids
  • Time to Results:
    1 h 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ANTIA313283-96
  • Format:
    Pre-coated
  • Target protein:
    NADH2
  • Detektionsmetode:
    Colorimetric
  • Prøvevolumen:
    40 μl
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO

Specifikationer