ABY Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Catalog # AATB67017
Supplier: AAT Bioquest
New Product
Specifications
- Rammefarve:hvid
- Ant. brønde:96
- Profil:Lav
- Skørt:Semi-skørt
- Brøndfarve:hvid
- Beskrivelse:ABY Dye qPCR calibration plate
- Stregkodet:Yes
- Pakn:1
Specifications
About this item
The ABY dye qPCR calibration plate is designed for use with the 7500 real-time PCR system featuring a fast 96-well block.
- The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
- This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
Caution: Avoid freeze-thawing plates. It is not recommended to use the plates more than 3 times. Plates can be stored for up to 12 months. Freeze (<−15 °C), and minimise light exposure.