Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Rat SDCCAG3 ELISA Kit

Rat SDCCAG3 ELISA Kit

Rat SDCCAG3 ELISA kit, 96 Tests
Katalog # ANTIA303725-96
Leverandør:   Antibodies.com
Rat SDCCAG3 ELISA Kit
Katalog # ANTIA303725-96
Leverandør:  Antibodies.com
CAS nummer:  
Rat SDCCAG3 ELISA Kit
Rat SDCCAG3 ELISA Kit

Rat SDCCAG3 ELISA kit, 96 Tests

CAS nummer:  
Price
4.890,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Rat SDCCAG3 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat SDCCAG3 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Rat SDCCAG3 ELISA kit (A303725) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat SDCCAG3 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for SDCCAG3 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SDCCAG3 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-SDCCAG3 Antibody, which binds the captured SDCCAG3 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of SDCCAG3 captured in each well. The concentration of SDCCAG3 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifikationer

Specifikationer

  • Pakn:
    96 Tests
  • Beskrivelse:
    Rat SDCCAG3 ELISA kit
  • Sensitivitet:
    18,75 pg/ml
  • Assay varighed:
    Multiple steps
  • Primary antibody reactivity:
    Rat
  • Detektionsområde:
    31,25 - 2000 pg/ml
  • Shelf life:
    Store for 6 months at 4 °C
  • Prøvetype:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Time to Results:
    4 h 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ANTIA303725-96
  • Format:
    Pre-coated
  • Krydsreaktivitet:
    Rat SDCCAG3 ELISA Kit exhibits high specificity and excellent specificity for the detection of rat SDCCAG3. No significant cross-reactivity or interference between SDCCAG3 and analogues was observed.
  • Target protein:
    SDCCAG3
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.