Mouse Prss29 ELISA Kit
Product Details & Documents
Mouse Prss29 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Prss29 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 0,156 to 10 ng/ml
- Sensitivity: 0,094 ng/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse Prss29 ELISA kit (A75763) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Prss29 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Prss29 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Prss29 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Prss29 Antibody, which binds the captured Prss29 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Prss29 captured in each well. The concentration of Prss29 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Bestillingsoplysninger: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Specifikationer
Specifikationer
- Pakn:96 Tests
- Beskrivelse:Mouse Prss29 ELISA kit
- Assay princip:Quantitative
- Sensitivitet:0,094 ng/ml
- Assay varighed:Multiple steps
- Primary antibody reactivity:Mouse
- Detektionsområde:0,156 - 10 ng/ml
- Shelf life:Store for 6 months at 4 °C
- Prøvetype:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:4 h 30 min
- Analysetype:Sandwich
- Cat. No.:ANTIA75763-96
- Format:Pre-coated
- Krydsreaktivitet:Mouse Prss29 ELISA kit exhibits high specificity and excellent specificity for the detection of mouse Prss29. No significant cross-reactivity or interference between Prss29 and analogues was observed.
- Target protein:Prss29
- Detektionsmetode:Colorimetric
- Prøvevolumen:100 μl
- Opbevaringstemperatur:4 °C
- Regulatory status:RUO



