Analytical Standard, Nicarbazin, Reference Material
Analytical standard, nicarbazin, reference material, 100 mg, 100 mg
Katalog # ACSDSB18251.100MG
Leverandør: CPACHEM
Analytical Standard, Nicarbazin, Reference Material
Katalog # ACSDSB18251.100MG
Leverandør: CPACHEM
CAS nummer:
Gå til:
Product Details & Documents
Bestillingsoplysninger: Shelf life on ship date: 24 months - store in a refrigerator at temperatures between 2 to 8 °C.
Specifikationer
Specifikationer
- Pakn:100 mg
- Beskrivelse:Analytical standard, nicarbazin, reference material
- Volumen:100 mg
- Cat. No.:ACSDSB18251.100MG
- CAS-nr:330-95-0
- GHS signal:Advarsel