Orders LinkContactUsLinkComponent
Hydrogen peroxide test kit, CHEMets®
Hydrogen peroxide HR CHEMets® test kit, 5 - 50 ppm, 1 KIT
Catalog # CHETK-5520C
Supplier:   CHEMetrics
Hydrogen peroxide test kit, CHEMets®
Catalog # CHETK-5520C
Supplier:  CHEMetrics
CAS Number:  
Hydrogen peroxide test kit, CHEMets®
Hydrogen peroxide HR CHEMets® test kit, 5 - 50 ppm, 1 KIT
CAS Number:  
Price
1.960,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
HR CHEMets® is the new way to test at high ranges and replaces the now discontinued VACUettes product line.
 : Test kits (5-50 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): HR CHEMets® refill, flat comparator, 25 ml sample cup, 10 ml syringe, and instructions.
Specifications
Specifications
  • Pakn:
    1 KIT
  • Beskrivelse:
    Hydrogen peroxide HR CHEMets® test kit
  • Område:
    5 - 50 ppm
Specifications
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Område
    Kemisk metode
    Availability
    Price
  • CHETR-5510
    CHEMets refill pack, 30 ampoules
    0 - 0,8; 1 - 10 ppm
    Ferric thiocyanate
    Log in for availability
    1 * 1 Each - 491,00 kr