Specifications
- Antigen name:CD14
- Clonality:Monoclonal
- Clone:M5E2
- Konjugation:Unconjugated
- Vært:Mouse
- Isotype:IgG2a kappa
- Reactivity:Dog,Squirrel monkey,Capuchin monkey,Pig,Sheep,Cynomolgus monkey,Common marmoset,Rhesus monkey,Cotton-topped tamarin,Macaque,Chimpanzee,Human,Bovine
- Gen ID:929
- Pakn:100 µG
Specifications
About this item
Mouse monoclonal IgG2a antibody against human, rhesus, cynomolgus CD14, unconjugated.
- Monocytes
The M5E2 antibody reacts with CD14, an ~53 to 55 kDa GPI-anchored transmembrane glycoprotein expressed at high levels on the surface of peripheral blood monocytes and macrophages, and at lower levels on granulocytes. An ~10-fold difference in expression levels between monocytes/macrophages and granulocytes makes CD14 a useful marker for distinguishing these cell populations. CD14 is also found on tissue macrophages, Langerhans cells and dendritic cells. CD14 functions as a high-affinity receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein and modulates LPS-dependent signal transduction during the immune response to gram-negative pathogens by acting as a co-receptor for TLR 4 and MD-2. This triggers activation of NF-kappa-B, cytokine secretion, and induction of the inflammatory response. Two soluble forms of CD14 have also been described (~48 and ~55 kDa). This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human Myeloid Positive Selection Kit (Catalog #18653) and EasySep™ Human CD14 Positive Selection Kit (Catalog #18058); partial blocking may be observed, as well as EasySep™ HLA Whole Blood CD33 Positive Selection Kit (Catalog #18287HLA), EasySep™ HLA Whole Blood Myeloid Positive Selection Kit (Catalog #18683HLA), and EasySep™ Human Buffy Coat CD14 Positive Selection Kit (Catalog #18088).
Type:
Antigen: CD14
Clonality: Monoclonal
Clone: M5E2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Capuchin monkey, Chimpanzee, Common marmoset, Cotton-topped tamarin, Bovine, Cynomolgus monkey, Dog, Human, Macaque, Pig, Rhesus monkey, Sheep, Squirrel monkey